siRNA Details
| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285511.1 | NP_001272440.1 | 8 | 1023 | ||
| NM_001285514.1 | NP_001272443.1 | 8 | 897 | |||
| NM_144910.2 | NP_659159.1 | 8 | 891 | |||
| NM_178854.4 | NP_849185.2 | 8 | 901 | |||
| GenBank mRNA | AK030112.1 | - | 8 | 899 | ||
| AK151728.1 | - | 8 | 1026 | |||
| BC018506.1 | - | 8 | 891 | 3598248 | ||
| BC023923.1 | - | 4 | 413 | |||

Cnot6l
Genomic Map