siRNA Details
| Gene Symbol: Tmem150c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182841.1 | NP_878261.1 | 6 | 699 | ||
| GenBank mRNA | AK044570.1 | - | 6 | 460 | ||
| AK044702.1 | - | 5 | 354 | |||
| AK166646.1 | - | 6 | 518 | |||
| BC028943.1 | - | 1 | 20 | |||
| BC055321.1 | - | 6 | 699 | 6419443 | ||
| BC062109.1 | - | 6 | 479 | |||

Tmem150c
Genomic Map