siRNA Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115010.1 | NP_001108482.1 | 13 | 2470 | ||
| NM_172714.4 | NP_766302.2 | 13 | 2338 | |||
| GenBank mRNA | AK029288.1 | - | 14 | 1784 | ||
| AK030894.1 | - | 13 | 2472 | |||
| AK134320.1 | - | 13 | 2336 | |||
| AK220225.1 | - | 13 | 2333 | |||
| BC125580.1 | - | 11 | 1465 | |||
| BC145025.1 | - | 11 | 1465 | 9053573 | ||

Lin54
Genomic Map