siRNA Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172715.3 | NP_766303.1 | 12 | 1461 | ||
| GenBank mRNA | AK030171.1 | - | 13 | 1646 | ||
| AK138410.1 | - | 11 | 1621 | |||
| AK160261.1 | - | 12 | 1406 | |||
| BC138228.1 | - | 11 | 1214 | 8861247 | ||

Agpat9
Genomic Map