siRNA Details
| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 867 | ||
| NM_178701.3 | NP_848816.3 | 3 | 857 | |||
| GenBank mRNA | AK046249.1 | - | 3 | 866 | ||
| AK050400.1 | - | 3 | 827 | |||
| AK080722.1 | - | 3 | 801 | |||
| AK137459.1 | - | 3 | 868 | |||
| AK167985.1 | - | 2 | 575 | |||
| BC037717.1 | - | 3 | 838 | 4239389 | ||

Lrrc8d
Genomic Map