siRNA Details
| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 2546 | ||
| NM_178701.3 | NP_848816.3 | 3 | 2536 | |||
| GenBank mRNA | AK046249.1 | - | 3 | 2545 | ||
| AK050400.1 | - | 3 | 2506 | |||
| AK080722.1 | - | 3 | 2480 | |||
| AK137459.1 | - | 3 | 2547 | |||
| AK167985.1 | - | 2 | 2254 | |||
| AK213350.1 | - | 1 | 100 | |||
| BC014756.1 | - | 1 | 1570 | |||
| BC037717.1 | - | 3 | 2517 | 4239389 | ||

Lrrc8d
Genomic Map