siRNA Details
| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289577.1 | NP_001276506.1 | 7 | 777 | ||
| NM_001289578.1 | NP_001276507.1 | 6 | 561 | |||
| NM_001289579.1 | NP_001276508.1 | 8 | 850 | |||
| NM_001289580.1 | NP_001276509.1 | 8 | 947 | |||
| NM_172717.4 | NP_766305.2 | 7 | 777 | |||
| GenBank mRNA | AK048930.1 | - | 8 | 927 | ||
| AK052473.1 | - | 7 | 732 | |||
| AK077629.1 | - | 7 | 749 | |||
| AK133154.1 | - | 8 | 832 | |||
| AK147892.1 | - | 6 | 536 | |||
| AK148436.1 | - | 7 | 748 | |||
| AK154038.1 | - | 7 | 759 | |||
| AK155295.1 | - | 7 | 749 | |||
| AK155525.1 | - | 7 | 749 | |||
| AK169182.1 | - | 8 | 803 | |||
| AK169193.1 | - | 7 | 732 | |||
| AK196958.1 | - | 4 | 373 | |||
| BC049792.1 | - | 7 | 731 | 6510480 | ||

Chfr
Genomic Map