siRNA Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198109.4 | NP_932777.2 | 12 | 1085 | ||
| GenBank mRNA | AB099287.1 | - | 12 | 1120 | ||
| AK089308.1 | - | 12 | 1089 | |||
| AK155557.1 | - | 12 | 1110 | |||
| AK165380.1 | - | 11 | 1050 | |||
| AK170787.1 | - | 11 | 1010 | |||
| AK171210.1 | - | 12 | 1134 | |||
| AK171556.1 | - | 12 | 1100 | |||
| AK173144.1 | - | 11 | 1041 | |||
| BC046529.1 | - | 6 | 567 | |||

Ssh1
Genomic Map