siRNA Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198109.4 | NP_932777.2 | 2 | 113 | ||
| GenBank mRNA | AB099287.1 | - | 2 | 148 | ||
| AK089308.1 | - | 2 | 117 | |||
| AK155557.1 | - | 2 | 138 | |||
| AK165380.1 | - | 1 | 76 | |||
| AK170787.1 | - | 2 | 103 | |||
| AK171210.1 | - | 2 | 162 | |||
| AK171556.1 | - | 2 | 128 | |||
| AK173144.1 | - | 2 | 135 | |||

Ssh1
Genomic Map