siRNA Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198109.4 | NP_932777.2 | 13 | 1347 | ||
| GenBank mRNA | AB099287.1 | - | 13 | 1382 | ||
| AK089308.1 | - | 13 | 1351 | |||
| AK155557.1 | - | 13 | 1372 | |||
| AK165380.1 | - | 12 | 1312 | |||
| AK170787.1 | - | 12 | 1272 | |||
| AK171210.1 | - | 13 | 1396 | |||
| AK171556.1 | - | 13 | 1362 | |||
| AK173144.1 | - | 12 | 1303 | |||
| AK187982.1 | - | 1 | 44 | |||
| BC046529.1 | - | 7 | 829 | |||

Ssh1
Genomic Map