siRNA Details
| Gene Symbol: Fbxw8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172721.2 | NP_766309.2 | 6 | 976 | ||
| GenBank mRNA | AK031815.1 | - | 6 | 970 | ||
| AK034447.1 | - | 6 | 904 | |||
| AK047695.1 | - | 6 | 944 | |||
| AK047756.1 | - | 6 | 918 | |||
| AK053794.1 | - | 6 | 977 | |||
| AK134291.1 | - | 6 | 983 | |||
| BC009095.1 | - | 6 | 929 | 3158218 | ||

Fbxw8
Genomic Map