siRNA Details
| Gene Symbol: Trafd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163470.1 | NP_001156942.1 | 8 | 1066 | ||
| NM_172275.2 | NP_758479.2 | 8 | 1066 | |||
| GenBank mRNA | AK150038.1 | - | 8 | 1035 | ||
| BC023072.1 | - | 3 | 998 | |||
| BC038396.1 | - | 8 | 1022 | 3987227 | ||

Trafd1
Genomic Map