siRNA Details
| Gene Symbol: B3gnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198611.2 | NP_941013.2 | 1 | 606 | ||
| GenBank mRNA | AY037786.1 | - | 1 | 607 | ||
| BC114987.1 | - | 1 | 561 | 40056240 | ||
| BC115755.1 | - | 1 | 561 | 40056241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt4| Gene Symbol: B3gnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198611.2 | NP_941013.2 | 1 | 606 | ||
| GenBank mRNA | AY037786.1 | - | 1 | 607 | ||
| BC114987.1 | - | 1 | 561 | 40056240 | ||
| BC115755.1 | - | 1 | 561 | 40056241 | ||