siRNA Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172277.2 | NP_758481.1 | 6, 7 | 769 | ||
| GenBank mRNA | AK045061.1 | - | 6, 7 | 762 | ||
| AK082871.1 | - | 6, 7 | 762 | |||
| AK148429.1 | - | 6, 7 | 762 | |||
| AK156817.1 | - | 6, 7 | 715 | |||
| AK161672.1 | - | 5, 6 | 703 | |||
| BC019142.1 | - | 5, 6 | 765 | |||
| BC037599.1 | - | 6, 7 | 795 | 5150754 | ||

Snx8
Genomic Map