siRNA Details
| Gene Symbol: Amz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173405.2 | NP_775581.1 | 4 | 784 | ||
| GenBank mRNA | AJ879912.1 | - | 2 | 425 | ||
| AK078332.1 | - | 4 | 784 | |||
| AK155136.1 | - | 671 | ||||
| AK170176.1 | - | 3 | 758 | |||
| BC129841.1 | - | 2 | 567 | 40090727 | ||
| BC129842.1 | - | 2 | 519 | 40090729 | ||

Amz1
Genomic Map