siRNA Details
| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289588.1 | NP_001276517.1 | 4, 5 | 1604 | ||
| NM_178702.4 | NP_848817.2 | 4, 5 | 1604 | |||
| GenBank mRNA | AK038570.1 | - | 4, 5 | 1537 | ||
| AK052964.1 | - | 4, 5 | 1515 | |||
| AK083410.1 | - | 4, 5 | 1622 | |||
| AK154169.1 | - | 4, 5 | 1545 | |||
| AK162917.1 | - | 4, 5 | 1585 | |||
| AK173277.1 | - | 4, 5 | 1568 | |||
| BC056483.1 | - | 4, 5 | 1556 | 5696312 | ||

Radil
Genomic Map