siRNA Details
| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289588.1 | NP_001276517.1 | 2 | 232 | ||
| NM_178702.4 | NP_848817.2 | 2 | 232 | |||
| GenBank mRNA | AK038570.1 | - | 2 | 165 | ||
| AK052964.1 | - | 2 | 143 | |||
| AK083410.1 | - | 2 | 250 | |||
| AK154169.1 | - | 2 | 171 | |||
| AK162917.1 | - | 2 | 212 | |||
| AK163852.1 | - | 2 | 168 | |||
| AK173277.1 | - | 2 | 196 | |||
| BC056483.1 | - | 2 | 184 | 5696312 | ||
| BY201066.1 | - | 2 | 234 | |||

Radil
Genomic Map