siRNA Details
| Gene Symbol: Fbxl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033312.3 | NP_001028484.2 | 3 | 1709 | ||
| GenBank mRNA | AK049302.1 | - | 3 | 1663 | ||
| AK144757.1 | - | 3 | 1711 | |||
| AK147353.1 | - | 2 | 3477 | |||
| AK154354.1 | - | 3 | 1689 | |||

Fbxl18
Genomic Map