siRNA Details
| Gene Symbol: PLXNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025179.3 | NP_079455.3 | 8 | 2693 | ||
| XM_005273164.2 | XP_005273221.1 | 8 | 2757 | |||
| XM_005273165.3 | XP_005273222.1 | 8 | 2757 | |||
| GenBank mRNA | AB007932.1 | - | 8 | 2144 | ||
| BC132676.1 | - | 8 | 2048 | 40146698 | ||
| BC136530.1 | - | 8 | 2048 | 9020520 | ||
| X87831.2 | - | 6 | 598 | |||

PLXNA2
Genomic Map