siRNA Details
| Gene Symbol: E130309D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172726.4 | NP_766314.3 | 4 | 918 | ||
| GenBank mRNA | AK041667.1 | - | 4 | 864 | ||
| AK050733.1 | - | 4 | 832 | |||
| AK053801.1 | - | 4 | 919 | |||
| AK054218.1 | - | 4 | 619 | |||
| AK087531.1 | - | 4 | 919 | |||
| AK172614.1 | - | 4 | 865 | |||
| AK198635.1 | - | 2 | 105 | |||
| BC058940.1 | - | 3 | 2997 | |||
| BC079853.1 | - | 4 | 852 | 5716912 | ||
| BC082321.1 | - | 4 | 662 | 6411746 | ||

E130309D02Rik
Genomic Map