siRNA Details
| Gene Symbol: Uspl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013378.2 | NP_001013396.2 | 4 | 573 | ||
| NM_001115149.1 | NP_001108621.1 | 4 | 573 | |||
| NM_001115150.1 | NP_001108622.1 | 3 | 404 | |||
| NM_001115151.1 | NP_001108623.1 | 3 | 404 | |||
| NM_001115153.1 | NP_001108625.1 | 5 | 750 | |||
| NM_001286824.1 | NP_001273753.1 | 4 | 531 | |||
| GenBank mRNA | AK050969.1 | - | 4 | 557 | ||
| AK088787.1 | - | 3 | 270 | |||
| AK145409.1 | - | 5 | 648 | |||
| AK161654.1 | - | 3 | 251 | |||
| AK168981.1 | - | 4 | 425 | |||
| BB655282.1 | - | 4 | 552 | |||
| BC028849.1 | - | 4 | 467 | |||
| BC032291.1 | - | 1 | 2543 | |||
| BC086664.1 | - | 4 | 415 | |||
| BY298649.1 | - | 3 | 301 | |||

Uspl1
Genomic Map