siRNA Details
Gene Symbol: Gimap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146167.3 | NP_666279.1 | 2 | 570 | ||
GenBank mRNA | AK040029.1 | - | 2 | 529 | ||
BC026200.1 | - | 2 | 532 | 963503 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gimap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146167.3 | NP_666279.1 | 2 | 570 | ||
GenBank mRNA | AK040029.1 | - | 2 | 529 | ||
BC026200.1 | - | 2 | 532 | 963503 |