siRNA Details
| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168277.1 | NP_001161749.1 | 3, 4 | 453 | ||
| NM_173406.3 | NP_775582.2 | 3, 4 | 452 | |||
| GenBank mRNA | AK043767.1 | - | 3, 4 | 402 | ||
| AK050474.1 | - | 3, 4 | 424 | |||
| BC048577.1 | - | 3, 4 | 453 | 6706223 | ||

Jazf1
Genomic Map