siRNA Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153573.1 | NP_705801.1 | 4 | 672 | ||
| GenBank mRNA | AK081692.1 | - | 4 | 699 | ||
| AK163384.1 | - | 818 | ||||
| BC024846.1 | - | 1099 | ||||
| BC029109.1 | - | 4 | 672 | 3985764 | ||
| BC034313.1 | - | 5 | 787 | 5365046 | ||

Fkbp14
Genomic Map