siRNA Details
| Gene Symbol: PLXNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163257.1 | NP_001156729.1 | 36, 37 | 5958 | ||
| NM_005393.2 | NP_005384.2 | 35, 36 | 5792 | |||
| GenBank mRNA | AB033032.1 | - | 35, 36 | 5732 | ||
| AF149019.1 | - | 35, 36 | 5732 | |||
| AK292535.1 | - | 35, 36 | 5792 | |||
| AK295762.1 | - | 35, 36 | 5812 | |||
| AK295894.1 | - | 34, 35 | 4676 | |||

PLXNB3
Genomic Map