siRNA Details
| Gene Symbol: PLXNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163257.1 | NP_001156729.1 | 14, 15 | 2687 | ||
| NM_005393.2 | NP_005384.2 | 13, 14 | 2521 | |||
| GenBank mRNA | AB033032.1 | - | 13, 14 | 2461 | ||
| AF149019.1 | - | 13, 14 | 2461 | |||
| AK292535.1 | - | 13, 14 | 2521 | |||
| AK295762.1 | - | 13, 14 | 2541 | |||
| AK295849.1 | - | 10, 11 | 1211 | |||
| AK295894.1 | - | 12, 13 | 1405 | |||
| AK304705.1 | - | 13, 14 | 1586 | |||
| AK307894.1 | - | 14, 15 | 2664 | |||

PLXNB3
Genomic Map