siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033929.2 | NP_001029101.1 | 2, 3 | 323 | ||
| NM_178413.5 | NP_848500.3 | 3, 4 | 391 | |||
| GenBank mRNA | AK165463.1 | - | 2, 3 | 289 | ||
| BC025604.1 | - | 3, 4 | 354 | 5250319 | ||
| BC051244.1 | - | 2, 3 | 285 | 4975661 | ||
| CA491740.1 | - | 2, 3 | 356 | |||
| CB600010.1 | - | 3, 4 | 361 | |||
| CX223060.1 | - | 2, 3 | 323 | |||

Thnsl2
Genomic Map