siRNA Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243741.1 | NP_001230670.1 | 3 | 997 | ||
| NM_001243742.1 | NP_001230671.1 | 3 | 533 | |||
| NM_146170.4 | NP_666282.3 | 3 | 474 | |||
| GenBank mRNA | AK047127.1 | - | 3 | 475 | ||
| AK077101.1 | - | 300 | ||||
| AK077503.1 | - | 3 | 884 | |||
| AK193994.1 | - | 1 | 26 | |||
| BC025632.1 | - | 3 | 441 | 5321669 | ||
| BC026926.1 | - | 3 | 544 | 3586097 | ||
| BC031913.1 | - | 3 | 487 | 5376246 | ||

C87436
Genomic Map