siRNA Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145148.2 | NP_660130.2 | 22 | 2851 | ||
| GenBank mRNA | AB042027.1 | - | 22 | 2774 | ||
| AF327856.1 | - | 21 | 2474 | |||
| AF327857.1 | - | 20 | 2261 | |||
| AK031491.1 | - | 11 | 2037 | |||
| AK129262.1 | - | 21 | 2326 | |||
| AK132280.1 | - | 22 | 2715 | |||
| BC031369.1 | - | 6 | 965 | |||
| BC039783.1 | - | 1 | 281 | |||
| BC052390.1 | - | 22 | 2720 | 5718942 | ||
| BC058262.1 | - | 11 | 1674 | 6487449 | ||

Frmd4b
Genomic Map