siRNA Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145148.2 | NP_660130.2 | 17, 18 | 1924 | ||
| GenBank mRNA | AB042027.1 | - | 17, 18 | 1847 | ||
| AF327856.1 | - | 16, 17 | 1547 | |||
| AF327857.1 | - | 15, 16 | 1334 | |||
| AK031491.1 | - | 6, 7 | 1110 | |||
| AK129262.1 | - | 16, 17 | 1399 | |||
| AK132280.1 | - | 17, 18 | 1788 | |||
| BC031369.1 | - | 1, 2 | 38 | |||
| BC052390.1 | - | 17, 18 | 1793 | 5718942 | ||
| BC058262.1 | - | 6, 7 | 747 | 6487449 | ||

Frmd4b
Genomic Map