siRNA Details
Gene Symbol: Gxylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198612.2 | NP_941014.1 | 6 | 1135 | ||
GenBank mRNA | BC049816.1 | - | 6 | 1135 | 6509198 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gxylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198612.2 | NP_941014.1 | 6 | 1135 | ||
GenBank mRNA | BC049816.1 | - | 6 | 1135 | 6509198 |