siRNA Details
| Gene Symbol: Ppp4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182939.4 | NP_891984.1 | 3 | 503 | ||
| GenBank mRNA | AK082539.1 | - | 3 | 504 | ||
| AK084169.1 | - | 3 | 464 | |||
| BC056340.1 | - | 3 | 323 | 5703151 | ||
| BC062117.1 | - | 2 | 177 | |||
| BC098464.1 | - | 3 | 269 | 30093511 | ||
| BC110429.1 | - | 3 | 508 | 5350332 | ||
| BC112399.1 | - | 2 | 81 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106613, XM_006236962, CB605683, EV770270, BC167050.

Ppp4r2
Genomic Map