siRNA Details
| Gene Symbol: Zfp637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177684.2 | NP_808352.1 | 2, 3 | 159 | ||
| GenBank mRNA | AB221671.1 | - | 1, 2 | 62 | ||
| BC048843.1 | - | 2, 3 | 159 | 5031082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp637| Gene Symbol: Zfp637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177684.2 | NP_808352.1 | 2, 3 | 159 | ||
| GenBank mRNA | AB221671.1 | - | 1, 2 | 62 | ||
| BC048843.1 | - | 2, 3 | 159 | 5031082 | ||