siRNA Details
| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081112.1 | NP_001074581.1 | 29, 30 | 4338 | ||
| GenBank mRNA | BC008111.1 | - | 9, 10 | 1884 | ||
| BC056364.1 | - | 12, 13 | 2397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd26| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081112.1 | NP_001074581.1 | 29, 30 | 4338 | ||
| GenBank mRNA | BC008111.1 | - | 9, 10 | 1884 | ||
| BC056364.1 | - | 12, 13 | 2397 | |||