siRNA Details
| Gene Symbol: Clec2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153506.4 | NP_705726.3 | 5 | 628 | ||
| GenBank mRNA | BC051091.1 | - | 5 | 642 | 6529908 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2e| Gene Symbol: Clec2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153506.4 | NP_705726.3 | 5 | 628 | ||
| GenBank mRNA | BC051091.1 | - | 5 | 642 | 6529908 | |