siRNA Details
| Gene Symbol: Clec12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177686.4 | NP_808354.1 | 3 | 405 | ||
| GenBank mRNA | AK084335.1 | - | 3 | 363 | ||
| BC094904.1 | - | 3 | 380 | 30044572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec12a| Gene Symbol: Clec12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177686.4 | NP_808354.1 | 3 | 405 | ||
| GenBank mRNA | AK084335.1 | - | 3 | 363 | ||
| BC094904.1 | - | 3 | 380 | 30044572 | ||