siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205363.1 | NP_001192292.1 | 6 | 691 | ||
| NM_001205364.1 | NP_001192293.1 | 5 | 610 | |||
| NM_172732.3 | NP_766320.2 | 5 | 613 | |||
| GenBank mRNA | AK036399.1 | - | 6 | 692 | ||
| AK041288.1 | - | 5 | 613 | |||
| BC087955.1 | - | 6 | 690 | 30119997 | ||
| EU339277.1 | - | 5 | 520 | |||
| EU339278.1 | - | 6 | 598 | |||
| EU339279.1 | - | 5 | 517 | |||

Clec9a
Genomic Map