siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205363.1 | NP_001192292.1 | 5 | 524 | ||
| NM_001205364.1 | NP_001192293.1 | 4 | 443 | |||
| NM_172732.3 | NP_766320.2 | 4 | 446 | |||
| GenBank mRNA | AK036399.1 | - | 5 | 525 | ||
| AK041288.1 | - | 4 | 446 | |||
| BC087955.1 | - | 5 | 523 | 30119997 | ||
| EU339277.1 | - | 4 | 353 | |||
| EU339278.1 | - | 5 | 431 | |||
| EU339279.1 | - | 4 | 350 | |||
| EU339280.1 | - | 4 | 353 | |||

Clec9a
Genomic Map