siRNA Details
| Gene Symbol: Gm156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014997.1 | NP_001014997.1 | 6 | 588 | ||
| GenBank mRNA | AB121775.1 | - | 6 | 588 | ||
| AK138706.1 | - | 7 | 1829 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm156| Gene Symbol: Gm156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014997.1 | NP_001014997.1 | 6 | 588 | ||
| GenBank mRNA | AB121775.1 | - | 6 | 588 | ||
| AK138706.1 | - | 7 | 1829 | |||