siRNA Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177687.3 | NP_808355.1 | 3 | 485 | ||
| GenBank mRNA | AK045467.1 | - | 3 | 486 | ||
| BC109358.1 | - | 3 | 357 | 40057562 | ||
| BC109359.1 | - | 3 | 373 | 40057564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebl2| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177687.3 | NP_808355.1 | 3 | 485 | ||
| GenBank mRNA | AK045467.1 | - | 3 | 486 | ||
| BC109358.1 | - | 3 | 357 | 40057562 | ||
| BC109359.1 | - | 3 | 373 | 40057564 | ||