siRNA Details
| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 1007 | ||
| GenBank mRNA | AK053755.1 | - | 1 | 932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2afj| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 1007 | ||
| GenBank mRNA | AK053755.1 | - | 1 | 932 | ||