siRNA Details
| Gene Symbol: Dera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172733.1 | NP_766321.1 | 8, 9 | 924 | ||
| GenBank mRNA | AK012558.1 | - | 8, 9 | 960 | ||
| AK075963.1 | - | 8, 9 | 949 | |||
| AK083298.1 | - | 8, 9 | 924 | |||
| AK145820.1 | - | 8, 9 | 960 | |||
| AK150273.1 | - | 8, 9 | 950 | |||
| AK150712.1 | - | 8, 9 | 920 | |||
| AK152462.1 | - | 8, 9 | 928 | |||
| AK168204.1 | - | 8, 9 | 960 | |||
| AK169418.1 | - | 8, 9 | 944 | |||
| AK199271.1 | - | 3 | 291 | |||
| BC016218.1 | - | 8, 9 | 930 | 4484163 | ||

Dera
Genomic Map