siRNA Details
| Gene Symbol: Klhl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081237.1 | NP_001074706.1 | 2 | 1356 | ||
| GenBank mRNA | AK048908.1 | - | 2 | 945 | ||
| AK052765.1 | - | 2 | 933 | |||
| AK083047.1 | - | 2 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl42| Gene Symbol: Klhl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081237.1 | NP_001074706.1 | 2 | 1356 | ||
| GenBank mRNA | AK048908.1 | - | 2 | 945 | ||
| AK052765.1 | - | 2 | 933 | |||
| AK083047.1 | - | 2 | 941 | |||