siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY250768.1 | - | 2 | 137 | ||
| CK621632.1 | - | 2 | 101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc136| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY250768.1 | - | 2 | 137 | ||
| CK621632.1 | - | 2 | 101 | |||