siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201378.1 | NP_001188307.1 | 6, 7 | 1028 | ||
| NM_145574.3 | NP_663549.3 | 6, 7 | 1028 | |||
| GenBank mRNA | BC006583.1 | - | 4, 5 | 616 | ||
| CX222432.1 | - | 3, 4 | 588 | |||
| CX564275.1 | - | 3, 4 | 533 | |||
| EU029664.1 | - | 6, 7 | 1028 | |||
| EU029665.1 | - | 6, 7 | 1028 | |||

Ccdc136
Genomic Map