siRNA Details
Gene Symbol: Ccdc136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145574.3 | NP_663549.3 | 15 | 3107 | ||
GenBank mRNA | AK160245.1 | - | 8 | 1834 | ||
EU029664.1 | - | 15 | 3107 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145574.3 | NP_663549.3 | 15 | 3107 | ||
GenBank mRNA | AK160245.1 | - | 8 | 1834 | ||
EU029664.1 | - | 15 | 3107 |