siRNA Details
| Gene Symbol: Zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172735.2 | NP_766323.1 | 9 | 1317 | ||
| GenBank mRNA | AJ537495.1 | - | 9 | 1309 | ||
| AK088527.1 | - | 9 | 1318 | |||
| AK146228.1 | - | 9 | 1332 | |||
| AK151694.1 | - | 9 | 1313 | |||
| AK167465.1 | - | 9 | 1318 | |||
| AK167992.1 | - | 9 | 1332 | |||
| AK168071.1 | - | 9 | 1327 | |||
| BC024560.1 | - | 3 | 333 | |||
| BC037445.1 | - | 9 | 1271 | |||
| BC050141.1 | - | 9 | 1310 | 30011212 | ||

Zc3hc1
Genomic Map