siRNA Details
| Gene Symbol: Shisa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290291.1 | NP_001277220.1 | 3 | 1051 | ||
| NM_172737.4 | NP_766325.3 | 3 | 1051 | |||
| GenBank mRNA | AK085127.1 | - | 2 | 273 | ||
| AK145016.1 | - | 2 | 336 | |||
| BY734785.1 | - | 2 | 336 | |||

Shisa7
Genomic Map