siRNA Details
| Gene Symbol: Zfp418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146179.2 | NP_666291.2 | 4 | 1074 | ||
| GenBank mRNA | AK039147.1 | - | 4 | 1036 | ||
| AK054428.1 | - | 4 | 1072 | |||
| BC024761.1 | - | 4 | 1054 | 5363178 | ||
| BC026490.1 | - | 4 | 1050 | 4504776 | ||

Zfp418
Genomic Map