siRNA Details
| Gene Symbol: Zscan22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001447.3 | NP_001001447.1 | 3 | 1664 | ||
| NM_001290436.1 | NP_001277365.1 | 3 | 1701 | |||
| NM_001290438.1 | NP_001277367.1 | 3 | 1598 | |||
| NM_001290440.1 | NP_001277369.1 | 2 | 1113 | |||
| NM_001290441.1 | NP_001277370.1 | 2 | 1179 | |||
| NM_001290442.1 | NP_001277371.1 | 2 | 1216 | |||
| GenBank mRNA | AK052559.1 | - | 3 | 1659 | ||
| AK078591.1 | - | 3 | 1568 | |||
| AK085207.1 | - | 3 | 1659 | |||
| AK163694.1 | - | 2 | 1190 | |||
| BC026676.1 | - | 2 | 1145 | 5053169 | ||

Zscan22
Genomic Map